Learn More
Thermo Scientific™ PEEK Capillary Tubing Available on GSA/VA Contract for Federal Government customers only.
Use these sample loops for your Thermo Scientific™ Rheodyne™ HPLC sample injectors. 
Manufacturer:  Thermo Scientific™ 3700710
 View more versions of this product
Catalog No. 03-050-393
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Use Thermo Scientific™ Rheodyne™ Sample Loops  for your Rheodyne HPLC sample injectors. They are available in stainless steel or biocompatible PEEK.
  • Broad chemical compatibility
  • Biocompatible
  • Easily cut to desired length
  • Appropriate for many HPLC applications
  • Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
Description & Specifications
Specifications
0.007 in. ID; 10cm length; Yellow
HPLC
Capillary Tubing
0.007 in. ID; 10cm length; Yellow