Thermo Scientific™ PEEK Capillary Tubing Available on GSA/VA Contract for Federal Government customers only.
Use these sample loops for your Thermo Scientific™ Rheodyne™ HPLC sample injectors. 
Manufacturer: Thermo Scientific™ 3702030
 View more versions of this product
Catalog No. 03-050-409
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With HPLC
    Type Capillary Tubing
    Item Description 0.020 in. ID; 30cm length; Orange
    Description 0.020 in. ID; 30cm length; Orange
    Use Thermo Scientific™ Rheodyne™ Sample Loops  for your Rheodyne HPLC sample injectors. They are available in stainless steel or biocompatible PEEK.
    • Broad chemical compatibility
    • Biocompatible
    • Easily cut to desired length
    • Appropriate for many HPLC applications
    • Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
  • Description & Specifications
Related Products
Product Content Feedback