Thermo Scientific™ PEEK Sleeves for Fused Silica HPLC Capillary Tubing Available on GSA/VA Contract for Federal Government customers only.
Make capillary tubing connections with Thermo Scientific™ PEEK Sleeves for Fused Silica Capillary Tubing.
Manufacturer: Thermo Scientific™ F230
 View more versions of this product
Catalog No. 03-052-430
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With Fused Silica Capillary Tubing
    Type Capillary Tubing Sleeve
    Item Description 0.015 in. ID; Orange
    Description 0.015 in. ID; Orange
    Use Thermo Scientific™ PEEK Sleeves for Fused Silica Capillary Tubing to connect capillary tubing to ports designed to receive 1/16" O.D. tubing. Chemically inert and flexible, they are capable of withstanding high pressures.
  • Description & Specifications
Related Products
Product Content Feedback