Learn More
Thermo Scientific™ PEEK Sleeves for Fused Silica HPLC Capillary Tubing
Make capillary tubing connections with Thermo Scientific™ PEEK Sleeves for Fused Silica Capillary Tubing.
$14.82
Specifications
Type Capillary Tubing Sleeve
For Use With Fused Silica Capillary Tubing
View More Specs
Products 6
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
03-052-429
View Documents
Thermo Scientific™
F229
0.012 in. ID; Natural Each for $14.82
Add to cart
03-052-430
View Documents
Thermo Scientific™
F230
0.015 in. ID; Orange Each for $14.82
Add to cart
03-052-427
View Documents
Thermo Scientific™
F227
0.008 in. ID; Yellow This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
03-052-428
View Documents
Thermo Scientific™
F228
0.010 in. ID; Blµe This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
03-052-431
View Documents
Thermo Scientific™
F231
0.021 in. ID; Natural This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
03-052-432
View Documents
Thermo Scientific™
F232
0.030 in.ID; Natural This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Use Thermo Scientific™ PEEK Sleeves for Fused Silica Capillary Tubing to connect capillary tubing to ports designed to receive 1/16 in. O.D. tubing. Chemically inert and flexible, they are capable of withstanding high pressures.
Specifications
Specifications
Capillary Tubing Sleeve
Fused Silica Capillary Tubing
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.