Learn More
MilliporeSigma™ Supelco™ PEEKsil™ Tubing
Fused silica tubing sheathed in PEEK polymer, with an effective outer diameter
Supplier: MilliporeSigma™ Supelco™ 51333U
Description
- PEEK sheating is mechanically strong and has ideal characteristics for sealing with metal or polymer fittings
- Fused silica core provides a consistent and rigid fluid pathway with very tight tolerances and industry-accepted chemical properties
- Extremely low absorption characteristics, especially when compared to stainless steel
- Excellent chemical compatibility
- Note: Do not cut this tubing - it should be used as its precut lengths because of permanent damage caused by conventional cutters
Specifications
Tubing | |
HPLC | |
1/16 in. | |
1.6 mm | |
Polymer-sheathed Fused Silica |
Orange | |
0.001 in. | |
25 μm | |
10 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
PEEKSil is a trademark of SGE Analytical Science Pty Ltd.