Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pefabloc™ SC, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Pefabloc(R) SC is a trypsin inhibitor, which can be used in the synthesis of its arginine-like analogue, 4-(2-guanidinoethyl)benzenesulfonyl fluoride.

$118.70 - $784.50

Specifications

Product Line Pefabloc
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Linear Formula C8H10 FNO2 S · HCl
Physical Form Powder or Crystals
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-3882
View Documents
MilliporeSigma Supelco
76307100MG
100 mg
Each for $118.70
Add to cart
 
11-101-3883
View Documents
MilliporeSigma Supelco
76307500MG
500 mg
Each for $451.00
Add to cart
 
11-101-3881
View Documents
MilliporeSigma Supelco
763071G
1 g
Each for $784.50
Add to cart
 
Description

Description

Pharmaceutical (small molecule)
Specifications

Specifications

Pefabloc
Analytical Standard
Powder or Crystals
≥95.0% (HPLC)
C8H10 FNO2 S · HCl
Safety and Handling

Safety and Handling

ShelfLife : 5 years

SDS
Product Certifications

Pefabloc is a registered trademark of Pentapharm

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit