Learn More
PEG3 Mouse anti-Human, Clone: OTI1B2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF801958
Catalog No. 50-166-5796
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
In human, ZIM2 and PEG3 are treated as two distinct genes though they share multiple 5 ft. exons and a common promoter and both genes are paternally expressed. Alternative splicing events connect their shared 5 ft. exons either with the remaining 4 exons unique to ZIM2, or with the remaining 2 exons unique to PEG3. In contrast, in other mammals ZIM2 does not undergo imprinting and, in mouse, cow, and likely other mammals as well, the ZIM2 and PEG3 genes do not share exons. Human PEG3 protein belongs to the Kruppel C2H2-type zinc finger protein family. PEG3 may play a role in cell proliferation and p53-mediated apoptosis. PEG3 has also shown tumor suppressor activity and tumorigenesis in glioma and ovarian cells. Alternative splicing of this PEG3 gene results in multiple transcript variants encoding distinct isoforms.
Specifications
Specifications
PEG3
Monoclonal
Unconjugated
PEG3
Lyophilized
PEG3
Human recombit protein fragment corresponding to amino acids 1-364 of human PEG3 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1B2
PBS with 8% trehalose and no preservative; pH 7.3
Q9GZU2
PW1, ZKSCAN22, ZNF904, ZSCAN24
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5178