Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEG3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551298

Catalog No. PA551298


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total PEG3 protein.

In human, ZIM2 and PEG3 are treated as two distinct genes though they share multiple 5' exons and a common promoter and both genes are paternally expressed. Alternative splicing events connect their shared 5' exons either with the remaining 4 exons unique to ZIM2, or with the remaining 2 exons unique to PEG3. In contrast, in other mammals ZIM2 does not undergo imprinting and, in mouse, cow, and likely other mammals as well, the ZIM2 and PEG3 genes do not share exons. Human PEG3 protein belongs to the Kruppel C2H2-type zinc finger protein family. PEG3 may play a role in cell proliferation and p53-mediated apoptosis. PEG3 has also shown tumor suppressor activity and tumorigenesis in glioma and ovarian cells. Alternative splicing of this PEG3 gene results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PEG3
Polyclonal
Unconjugated
PEG3
AL022617; ASF-1; End4; Gcap4; granule cell antiserum positive 4; KIAA0287; Kruppel-type zinc finger protein; mKIAA0287; paternally expressed 3; paternally expressed gene 3; paternally-expressed gene 3 protein; PEG3; Pw1; Zfp102; zinc finger and SCAN domain-containing protein 24; ZKSCAN22; ZNF904; ZSCAN24
Rabbit
Antigen affinity chromatography
RUO
5178
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9GZU2
PEG3
Synthetic peptide of human PEG3.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.