Learn More
Aviva Systems Biology Corp PELI3 Antibody - N-terminal region (ARP54532_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PELI3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PELI3. Alternate names: MGC35521ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61237