Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pellino 3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548582

Catalog No. PA548582

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Pellino 1 is the mammalian homologue of Pellino, a Drosophila Pelle-interacting protein. Pelle is a serine-threonine kinase through which Toll signaling can occur. Pellino 1 can activate c-Jun, Elk-1 and NF-kB, whereas Pellino 3 can only activate c-Jun and Elk-1(2).
Specifications

Specifications

Pellino 3
Polyclonal
PBS with 0.09% sodium azide
Q8N2H9
E3 ubiquitin-protein ligase pellino homolog 3, Pellino-3, 632-, PELI3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
246330
Flow Cytometry, Western Blot
Unconjugated
PELI3
Liquid
PELI3
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PELI3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit