Learn More
Pelican™ 0350 Cube Cases
Extra deep, unbreakable, watertight, airtight, dustproof, chemical resistant and corrosion proof. It is made from Ultra High Impact structural copolymer that makes it extremely strong and durable.
Manufacturer:  Pelican™ 0350
 View more versions of this product
Catalog No. 19-130-1085
Add to cart
Description
Description
  • Large 2-person fold down handles
  • Stainless steel hardware and padlock protectors
  • 3 level Pick N Pluck™ with convoluted lid foam
  • Watertight, crushproof, and dustproof
  • Open cell core with solid wall design - strong, light weight
  • Automatic Pressure Equalization Valve - balances interior pressure, keeps water out
  • O-ring seal
Certifications
IP67 / STANAG 4280 / Def Stan 81-41
Specifications
Specifications
50.8cm
32 lb.
56.9cm
50.8cm
21.25 in.
14.52kg
57.1cm
54cm
Double Throw latch system
22.5 in.
20 in.
Pelican Cube Case
Pick N' Pluck
Black
Pick N Pluck foam
20 in.
20 x 20 x 20 in. (50.8 x 50.8 x 50.8cm)
20 in.
50.8cm
22.43 in.
20 in.
50.8cm
High Impact Copolymer Polypropylene