Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican™ 0350 Cube Cases

Extra deep, unbreakable, watertight, airtight, dustproof, chemical resistant and corrosion proof. It is made from Ultra High Impact structural copolymer that makes it extremely strong and durable.

Manufacturer:  Pelican™ 0350NF

 View more versions of this product

Catalog No. 19-130-1086


Add to cart

Description

Description

  • Large 2-person fold down handles
  • Stainless steel hardware and padlock protectors
  • 3 level Pick N Pluck™ with convoluted lid foam
  • Watertight, crushproof, and dustproof
  • Open cell core with solid wall design - strong, light weight
  • Automatic Pressure Equalization Valve - balances interior pressure, keeps water out
  • O-ring seal

Certifications

IP67 / STANAG 4280 / Def Stan 81-41

Specifications

Specifications

14.52kg
Black
54cm
21.25 in.
50.8cm
50.8cm
20 in.
50.8cm
56.9cm
32 lb.
High Impact Copolymer Polypropylene
20 in.
22.5 in.
Pelican Cube Case
20 in.
Pick N' Pluck
Double Throw latch system
No foam
22.43 in.
20 in.
57.1cm
50.8cm
20 x 20 x 20 in. (50.8 x 50.8 x 50.8cm)
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit