Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican™ 2130 Mini Flasher LED Flashlight

Perfect for night-time bicyclists and joggers

Manufacturer:  Pelican™ 2130010245

Catalog No. 19-600-105


Add to cart

Description

Description

  • Hi-visibility LED flashlight that can be seen over half a mile away
  • Just twist to turn on
  • Powered by 2 L1154 1.2V alkaline coin cells that last over 100 hours before replacing
  • Submersible- 1m for 30 minutes

Specifications

Specifications

2.4V
ANSI/NEMA FL 1
Yellow
2 L1154 Alkaline
LED
0.01kg
Perfect for night-time bicyclists and joggers
Polycarbonate Resin
2130 Mini Flasher LED Flashlight
0.5 oz.
4.4cm
1.73 in.
2 L1154 alkaline batteries
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit