Learn More
Pelican™ 7060 LED Rechargeable Flashlight
Ideal tactical/patrol flashlight
Manufacturer:  Pelican™ 7060 LED
Catalog No. 19-168-762
Add to cart
Includes: 3.7V, 2200mAh battery; charging unit; 110V transformer; holster.
Description
Description
  • 2.25-hour burn time on high, 15 hours on low
  • Employs new dual-switch technology—allows light to be activated by either a body-mounted patrol switch or a tail-mounted tactical switch
  • Dual switches, which include a momentary mode, allow user to grip light in several positions and still control activation
  • Handle features grenade grip with no-slip pattern
  • Uses lightweight, rechargeable lithium-ion batteries for maximum brightness
Order Info
Length: 8.6 in. (22cm). Weight (with batteries): 10.4 oz. (295g).
Specifications
Specifications
7060 LED Flashlight
0.29kg
Lithium Ion
Ideal tactical or patrol flashlight, uses lightweight, rechargeable lithium-ion batteries for maximum brightness
8.65 in.
14768 cd
164lm
3.7V
10.4 oz.
2 hr. 15 min.
ANSI/NEMA FL 1
3.7V, 2200mAh battery, charging unit, 110V transformer, and holster
22cm
LED
Xenoy™
Black