Pelican™ 9460 Remote Area Lighting System

Convenient and ecologically responsible

Manufacturer: Pelican™ 9460000110

 View more versions of this product

Catalog No. 19-502-002

  Check Availability
Add to Cart

Alert

The Pelican Products RALS models 9460 and 9470 contain sealed lead acid batteries that can generate hydrogen gas when charging. It is imperative that these products are charged, as instructed, with the case lid open to allow any gases to escape as charging with the lid closed poses a significant hazard. Failure to abide by this instruction could result in injury and/or damage to property.


    Specifications

    Battery Run Time 7 hr.
    Battery Life 500 Cycles
    Battery Rechargeable
    Certifications/Compliance CE, RoHs
    For Use With (Application) Provides convenient and ecologically responsible alternatives to wasteful generator powered lights
    Height (English) 9 in.
    Height (Metric) 22.9cm
    Includes Thermally efficient light engine, 2 light heads, multi-stage charger, telescopic handle, rugged wheels
    Lamp Life 50000 hr.
    Light Source High flux LED
    Voltage 12V
    Width (English) 13.81 in.
    Width (Metric) 35.1cm
    Color Black
    Product Type 9460 Remote Area Lighting System
    Weight (English) 55 lb.
    Weight (Metric) 24.95kg

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit