Learn More
Pelican Big Ed Flashlight
Rugged, right-angle flashlight can be carried easily in the hand, worn on a shoulder strap, or clipped to turnout gear
$4.66
Specifications
Certifications/Compliance | UL, Class I, Division 2, Groups A, B, C and D, Temperature Rating T2A |
---|---|
Material | ABS/Polycarbonate Shroud |
Warranty | Lifetime |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
17-985-408A
|
Pelican
1901 |
Fast Charger |
Each for $4.66
|
|
Description
Choose from models powered by disposable “C”-cell alkaline (not included) or rechargeable NiCd batteries. Maximum candlepower varies between lights using different power sources.
- Unbreakable, water-resistant flashlight body is constructed from chemical-resistant resin covered in an ABS/polycarbonate shroud
- Unique push-button battery ejection system allows user to change out battery pack without tools
- Xenon lamp produces an exceptionally bright, long-distance beam
Not for diving applications.
Order Info
Weight: 20.5 oz. (0.58kg)
Compliance
UL listed for use in hazardous locations classified as Class I, Division 2, Groups A, B, C and D, Temperature Rating T2A.
Warranty and Services
Lifetime warranty
Specifications
UL, Class I, Division 2, Groups A, B, C and D, Temperature Rating T2A | |
ABS/Polycarbonate Shroud | |
Lifetime |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.