Pelican™ Helmet Light Holders

Locking cams and swivel settings allow light to be angled up or down and horizontally

Manufacturer: Pelican™ 750

 View more versions of this product

Catalog No. 19-816-498

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance Navy approved
    Color Navy Blue
    Material Stainless Steel
    Product Type Helmet Light Holder
    For Use With (Equipment) Helmet and hard hats for hands-free use of 2300, 2350, 2400 and 2450 flashlights

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit