Learn More
Pelican™ L4™ 1830 LED Lights
Lightweight, slim-profile design fits in pocket
Manufacturer:  Pelican™ 1830010110
 View more versions of this product
Catalog No. 19-150-0096
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1830 LED Light
0.03kg
Alkaline
Lightweight, slim-profile design fits in pocket
10000 hr.
15.7cm
LED
Xenoy™ Polycarbonate Resin
Black
1.2 oz.
32 hr.
FM, Class I, Division 1 hazardous locations, CE
Three AAAA batteries
6.2 in.
44 cd
5lm
4.5V