Pelican™ L4™ 1830 LED Lights

Lightweight, slim-profile design fits in pocket

Manufacturer: Pelican™ 1830010245

 View more versions of this product

Catalog No. 19-150-0097

  Check Availability
Add to Cart

    Specifications

    Battery Run Time 32 hr.
    Battery Alkaline
    Certifications/Compliance FM, Class I, Division 1 hazardous locations, CE
    For Use With (Application) Lightweight, slim-profile design fits in pocket
    Includes Three AAAA batteries
    Lamp Life 10000 hr.
    Length (English) 6.2 in.
    Length (Metric) 15.7cm
    Light Output 44 cd
    Light Source LED
    Lumen 5lm
    Material Xenoy™ Polycarbonate Resin
    Voltage 4.5V
    Color Yellow
    Product Type 1830 LED Light
    Weight (English) 1.2 oz.
    Weight (Metric) 0.03kg

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit