Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican™ M10 8040 Flashlight

High-pressure Xenon™-fired lamp module produces a powerful, bright, high intensity light

Manufacturer:  Pelican™ 8040 BLK

Catalog No. 19-816-500

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Built tough enough to withstand rough handling in the plant, on the beat or on patrol.

  • Tight, white prefocused beam
  • Polarity guard tray makes operating the flashlight impossible if cells are inverted; prevents dangerous outgassing
  • Runs 5.25 hours on four “C” alkaline batteries (not included)
  • Lightweight Xenoy polymer resin resists chemicals
  • Sure-grip sheath prevents slippage
  • Push button is easy to operate even when wearing thick gloves

Order Info

Length: 11.25 in. (28.6cm). Weight (with batteries): 8.4 oz. (239g).

Certifications

CSA and FM listed for certain Class 1, Division 2 hazardous locations.

Specifications

Specifications

18.4 oz.
6V
Black
5 hr. 15 min.
CSA, FM, Class 1, Division 2 hazardous locations
4 C alkaline batteries
28.6cm
Xenon
Xenoy™ Polymer Resin
0.52kg
4.8w
M10 8040 Flashlight
4 C Alkaline
High-pressure xenon™-fired lamp module produces a powerful, bright, high intensity light, built tough enough to withstand rough handling in the plant, on the beat or on patrol
11.25 in.
9820 cd
67lm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit