Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican™ MityLite™ Flashlights

Catalog No. p-119772
Click to view available options
Color:
Yellow
Length (English):
4.09 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Beam cuts through thick smoke, fog, and dust

  • 1.5 hours burn time
  • High-intensity Xenon™ Laser Spot™ lamp produces a 11 Lumens beam
  • Xenoy polymer resin body with twist-on shroud is submersible and IPX7 rating

Order Info

Length: 4.1 in. (10.4cm). Weight (with batteries): 1.6 oz. (45.4g). Two AAA alkaline batteries.

Certifications

CSA approved for certain Class I, Division 2 hazardous locations; CE, IEC, and ETL marked.

Specifications

Gas Fill Xenon
Voltage 3 V
Weight (English) 1.6 oz.
Weight (Metric) 0.05 kg
Battery Run Time 1.5 hr.
Battery Type Alkaline
Certifications/Compliance CSA, CE, IEC, and ETL
Color Yellow
For Use With (Application) Uses a hi-intensity xenon lamp that produces a powerful, bright beam, beam cuts through thick smoke, fog, and dust
Includes Two AAA alkaline batteries
Length (English) 4.09 in.
Length (Metric) 10.4 cm
Lumen 11 lm
Material Polymer Resin
Product Type Pocket Light
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.