Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican™ Magnum Flashlight

Focused Xenon Laser Spot™ beam is 600% brighter than ordinary flashlights

Manufacturer:  Pelican™ 2300CYLW

 View more versions of this product

Catalog No. 19-035-429


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

4.2 hr.
Shirt clip, two AA batteries
5.6 in.
8lm
1.8w
MityLite™ Magnum Flashlight
FM and CSA
20 hr.
13cm
Polycarbonate Resin
Yellow
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit