Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican™ SabreLite™ 2010 Recoil LED™ Flashlights

Produce a white collimated beam as bright as incandescent light

Manufacturer:  Pelican™ 2010PL

 View more versions of this product

Catalog No. 19-412-643


Add to cart

Description

Description

  • 32 hours of burn time
  • EXL body
  • Twist ON/OFF lens shroud eliminates accidental discharge

Order Info

Length: 8.1 in. (20.6cm). Weight (with batteries): 13.4 oz. (380g). Requires three C alkaline batteries.

Certifications

ETL listed for certain Class I, II, III, Division 1 hazardous locations. 1ECEx marked.

Specifications

Specifications

LED
Black
Three C alkaline batteries, photo luminescent shroud
32 hr.
ABS Resin
20.6cm
ETL, Class I, II, III, division 1, 1ECEx
On/Off
Produce a white collimated beam as bright as incandescent light
13.4 oz.
8.11 in.
109lm
Flashlight w/photoluminescent shroud
10000 hr.
0.38kg
Alkaline
4.5V
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit