Learn More
Pelican™ StealthLite™ Flashlight and Rechargeable System
IPX7 rating
Manufacturer:  Pelican™ 2400CORG
 View more versions of this product
Catalog No. 19-086789
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Flashlight
0.2kg
Alkaline
The tough resin body is corrosion proof and resistant to extreme temperatures, xenon laser spot lamp module with white collimated beam one-hand safety switch and double shock mounted battery compartment
6.49 in.
Xenon
LEXAN™ EXL Resin
Orange
7.1 oz.
3.45 hr.
CSA, FM, Class I, Division 2, ETL, ANSI/NEMA FL 1
4 AA alkaline batteries
16.48cm
45lm
6V