Pelican™ Super SabreLite™ Flashlights

Suitable for potentially explosive and other hazardous environments

Manufacturer: Pelican™ 2000C BLK

 View more versions of this product

Catalog No. 17-985-407A

  Check Availability
Add to Cart

Required Accessories : Three “C” alkaline batteries (not included).

    Specifications

    Battery Run Time 5 hr.
    Battery 3 C Alkaline
    Certifications/Compliance UL, Class 1. Division 1, Group C and D, ANSI/NEMA FL 1
    For Use With (Application) Suitable for potentially explosive and other hazardous environments
    Includes Spring clip, ring, and lanyard
    Length (English) 7.79 in.
    Length (Metric) 19.8cm
    Light Source Xenon
    Lumen 53lm
    Material ABS Plastic Resin
    Voltage 4.5V
    Color Black
    Product Type Flashlight
    Weight (English) 12.8 oz.
    Weight (Metric) 0.36kg

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit