Learn More
Pelican™ VersaBrite™ 2250 Flashlight
Manufacturer:  Pelican™ 2250C
Catalog No. 19-077577
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Multiple-position swivel head sealed with an O-ring
  • With 7 Lumens wide-beam xenon LMX lamp module
  • Unbreakable, chemical-resistant Xenoy™ resin body
  • Available in nonglare black
  • Clips rotate 360degree and come with double-sided adhesive
Specifications
Specifications
2 AA Alkaline
2 AA batteries, head strap, hook/loop, magnet and night vision discs
9.8cm
7
3V
Safety
ANSI/NEMA FL 1, Class I, Div. 2, Gp ABCD, T3C, Class ll; Div. 2; Gp G -35, Class I, Div. 2
3.87 in.
Xenon
Polycarbonate Resin
VersaBrite™ 2250 Deluxe Light Kit