Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican™ Watertight Equipment Cases

Open-cell core with solid wall design

Manufacturer:  Pelican™ 1560000110

 View more versions of this product

Catalog No. 19-160-872


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Watertight Equipment Case
1560
51.7cm
39.2cm
22.9cm
Double throw latches, padlock protectors
17 lb.
10.43 in.
Ultra High Impact Copolymer Polypropylene, Polyurethane
45.5cm
55.88cm
Black
20.37 in.
15.4 in.
9 in.
2 Level Pick N' Pluck
Rubber over-molded top and side handles
7.7kg
26.5cm
17 in.
22 in.
22 x 17 in. (55.88 x 45.5cm)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit