Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican™ 1200 Small Case

Lightweight cas is extremely strong, compact, and fully capable.

Manufacturer:  Pelican™ 1200ORG

 View more versions of this product

Catalog No. 19-816-449


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Watertight Equipment Case
1200
23.5cm
18.1cm
10.5cm
2.45 lb.
4.87 in.
Ultra High Impact Copolymer
24.6cm
27cm
Orange
9.25 in.
7.12 in.
4.12 in.
Pick N' Pluck
1.2kg
12.4cm
9.68 in.
10.62 in.
10.62 x 9.68 in. (27 x 24.6cm)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit