Learn More
Thermo Scientific™ Peltier Accessory (Air-Cooled) for GENESYS™ 10S series, BioMate™ 3S and Evolution™ 60S spectrophotometers
Thermo Scientific™ Air-Cooled Peltier Accessory for GENESYS 10S series, BioMate 3S and Evolution 60S spectrophotometers quickly specifies the temperature for the analysis and controls the rate of stirring with the external control module.
Manufacturer:  Thermo Scientific™ 222238100
 View more versions of this product
Catalog No. 14-386-461
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Description
Description
Thermo Scientific™ Peltier Accessory (Air-Cooled) for GENESYS™ 10S series, BioMate™ 3S and Evolution™ 60S Spectrophotometers quickly specifies the temperature for the analysis and controls the rate of stirring with the external control module. Temperature control and sample stirring are important for many UV-Visible analysis methods. The air-cooled Peltier accessory delivers superior performance in an easy-to-use configuration. It provides reliable temperature control from 20° to 60°C.
The Peltier control system provided high accuracy temperature control without the need to watch water levels, mix in anti-freeze or anti-bacterial, or plumb the accessory with hoses and clamps.
  • Control module features a large temperature display and graphical indication of the stirring speed
  • Module software allows you to change temperature or stirring speed quickly
  • Offers exceptional temperature stability and fast temperature transitions
  • Small temperature differential between set temperature of the accessory and temperature as measured inside a standard, 1cm cuvette
  • Less expensive than most recirculating liquid temperature controllers
  • No maintenance design
Ordering Alerts:
Sample stirring, requires stirring bars available separately (9423UV97030E). Designed for biologically relevant assays that require temperature control at 25°, 37°, 40°, and 50°C.
Recommended for:
Life science laboratory
Specifications
Specifications
Air-Colled Peltier System, single cell
Kit contains:
One sample compartment insert with thermostatted single cell holder
One replacement instrument lid with opening for air circulation for Peltier device
One external control unit
Temperature can be set from the control unit front panel or from VISIONlite software running both the Peltier accessory and the spectrophotometer from a connected Windows™ PC.
20°C to 60°C
GENESYS 10S Series, BioMate 3S, Evolution 60S spectrophotometers
Magnetic stirring of cell contents using included 2mm x 5mm Teflon-coated magnetic stir bar
± 0.1°C