Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G potassium salt, 100 mg/mL in distilled water, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
50mL
100mL
113-98-4
C16H17KN2O4S
372.48
MFCD00036193
IYNDLOXRXUOGIU-LQDWTQKMSA-M
23664709
CHEBI:7963
potassium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[K+]
113-98-4
C16H17KN2O4S
372.48
MFCD00036193
IYNDLOXRXUOGIU-LQDWTQKMSA-M
23664709
CHEBI:7963
potassium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[K+]
Packaging Plastic bottle
Physical Form Liquid
Quantity 100mL
Chemical Name or Material Penicillin G potassium salt