Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G Potassium Salt , MP Biomedical

Penicillin G Potassium Salt is a narrow spectrum antibiotic. It is the drug of choice for Streptococcus pneumoniae, groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridans group streptococci, and non-penicillinase producing staphylococcus.

$79.50

Specifications

Packaging Amber Glass Bottle
Purity 99.8
Color White
pH 5 to 7.5 (3% aq. soln.)
Melting Point 214°C to 217°C (literature)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP210054310 View Documents MP Biomedicals, IncSupplier Diversity Partner
0210054310
10mu Each for $79.50
Add to cart
 
Description

Description

  • Penicillin G potassium salt is used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutants
  • Active against gram-positive and gram-negative aerobic and anaerobic bacteria
  • Inhibits cell wall synthesis by binding to penicillin binding proteins which inhibit peptidoglycan chain cross-linking
  • Stable against hydrolysis by β-lactamases, such as penicillinases and cephalosporinases
Specifications

Specifications

Amber Glass Bottle
White
214°C to 217°C (literature)
99.8
5 to 7.5 (3% aq. soln.)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit