Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G sodium salt, 100mg/mL in distilled water, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
50mL
100mL
69-57-8
C16H17N2NaO4S
356.372
MFCD00069666
FCPVYOBCFFNJFS-LQDWTQKMSA-M
23668834
CHEBI:51765
sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]
69-57-8
C16H17N2NaO4S
356.372
MFCD00069666
FCPVYOBCFFNJFS-LQDWTQKMSA-M
23668834
CHEBI:51765
sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]
Packaging Plastic bottle
Physical Form Liquid
Quantity 100mL
Chemical Name or Material Penicillin G sodium salt