Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G sodium salt, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
69-57-8
C16H17N2NaO4S
356.372
FCPVYOBCFFNJFS-LQDWTQKMSA-M
23668834
CHEBI:51765
sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]
69-57-8
C16H17N2NaO4S
356.372
FCPVYOBCFFNJFS-LQDWTQKMSA-M
23668834
CHEBI:51765
sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]
Infrared Spectrum Authentic
Solubility Information Solubility in water: soluble.
Formula Weight 356.37
Percent Purity ≥97.5% (HPLC)
Specific Rotation +293° to +303° (20°C, 589nm) (c=2, H2O)
pH 5.0 to 7.5
Quantity 25g
Chemical Name or Material Penicillin G sodium salt

GHS H Statement May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. Wear eye protection/face protection.

Danger