Learn More
Penicillin/Streptomycin/Glutamine Mixture, Fisher BioReagents
Interferes with synthesis of bacterial cell wall. Inhibits protein synthesis. Interferes with bacterial cell division
Catalog No. BP296050
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • A sterile-filtered antibiotic solution used to prevent bacterial contamination of cell cultures
  • The two combined antibiotics prove effective against gram-positive and gram-negative bacteria
  • Penicillin interferes with the synthesis of bacterial cell wall and streptomycin inhibits protein synthesis in bacteria by binding to the 30S subunit of bacterial ribosome
  • Glutamine serves as an energy source for cells in culture
  • Intended for research use only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Penicillin/Streptomycin/Glutamine
Liquid
15, 4507
Pass Test
Penicillin/Streptomycin/Glutamine Mixture
3810-74-0, 69-57-8, 7647-14-5, 56-85-9, 7732-18-5
Plastic Bottle
50 mL
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes, respiratory system and skin. Substances known to cause developmental toxicity in humans. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:2
Flammability:0
Instability:0