Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin V potassium salt, 100 mg/mL in distilled water, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
50mL
100mL
132-98-9
C16H20KN2O5S
391.503
MFCD00051771
HWJJRHQIYDZECL-UDPGNSCCSA-N
131673927
(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;potassium
[HH].CC1(C(N2C(S1)C(C2=O)NC(=O)COC3=CC=CC=C3)C(=O)O)C.[K]
132-98-9
C16H20KN2O5S
391.503
MFCD00051771
HWJJRHQIYDZECL-UDPGNSCCSA-N
131673927
(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;potassium
[HH].CC1(C(N2C(S1)C(C2=O)NC(=O)COC3=CC=CC=C3)C(=O)O)C.[K]
Packaging Plastic bottle
Physical Form Liquid
Quantity 100mL
Chemical Name or Material Penicillin V potassium salt