Learn More
Bachem H-Ala-Ala-Ala-Ala-Ala-OH, 250mg, 10183-34-3, MFCD00080980 DFS Item
Manufacturer:  Bachem 4007939.0250
Pentaalanine with its strong tendency to form β-sheets may serve as a model for amyloid fibrillation and interactions of small molecules with Aβ.
Catalog No. 50-194-7438