Alfa Aesar™ Pentacarbonyliron, 99.5%

As a catalyst in organic reactions, and as carbonyl iron for high-frequency coils

Manufacturer: Alfa Aesar™ 31113A1

Quantity 1kg

 View more versions of this product

Catalog No. AA31113A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13463-40-6
    Linear Formula Fe(CO)5
    Formula Weight 195.9
    MDL Number MFCD00011001
    UN Number UN1994
    Merck Index 14,5099
    Synonym Iron pentacarbonyl; Iron carbonyl
    Chemical Name or Material Pentacarbonyliron

    Specifications

    Formula Weight 195.9
    Health Hazard 3 GHS P Statement
    P210-P280h-P304+P340-P310-P302+P352
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    MDL Number MFCD00011001
    UN Number UN1994
    Health Hazard 2 GHS H Statement
    H225-H300-H310-H330
    Highly flammable liquid and vapor.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Refractive Index 1.5196
    Synonym Iron pentacarbonyl; Iron carbonyl
    Health Hazard 1 Danger
    Color Red
    Boiling Point 103°C (200°C (dec.))
    Odor Pungent
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: IRON PENTACARBONYL
    Merck Index 14,5099
    Melting Point −20°C
    Flash Point −15° (5°F)
    Sensitivity Air & Light Sensitive
    Density 1.457g/mL
    TSCA TSCA
    Physical Form Liquid
    Linear Formula Fe(CO)5
    Solubility Information Soluble in nickel tetracarbonyl and organic solvents; Insoluble in water
    Quantity 1kg
    CAS 13463-40-6
    RTECS Number NO4900000
    Chemical Name or Material Pentacarbonyliron
    Assay 99.5%

    GHS H Statement
    H225-H300-H310-H330
    Highly flammable liquid and vapor.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.

    GHS P Statement
    P210-P280h-P304+P340-P310-P302+P352
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit