Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Pentachloroethane, 96%

Manufacturer:  Alfa Aesar™ 4447518

Quantity 50g

 View more versions of this product

Catalog No. AA4447518


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

76-01-7
CCl3CHCl2
MFCD00000846
1736845
Pentachloroethane
C2HCl5
202.3
UN1669
14,7108

Specifications

76-01-7
Liquid
CCl3CHCl2
MFCD00000846
1736845
1.5030
−29°C
50g
Pentachloroethane
1.68g/mL
C2HCl5
202.3
UN1669
14,7108
Colorless
162°C
Like chlorine
96%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H372-H351
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.

GHS P Statement
P260-P281-P264-P308+P313-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
Wash thoroughly after handling.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PENTACHLOROETHANE

RTECSNumber : KI6300000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit