Alfa Aesar™ Pentachloroethane, 96%

Manufacturer: Alfa Aesar™ 4447509

Quantity 10g

 View more versions of this product

Catalog No. AA4447509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76-01-7
    Molecular Formula C2HCl5
    Linear Formula CCl3CHCl2
    Formula Weight 202.3
    MDL Number MFCD00000846
    UN Number UN1669
    Beilstein 1736845
    Merck Index 14,7108
    Chemical Name or Material Pentachloroethane

    Specifications

    Formula Weight 202.3
    Health Hazard 3 GHS P Statement
    P260-P281-P264-P308+P313-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    Wash thoroughly after handling.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00000846
    Color Colorless
    Boiling Point 162°C
    Odor Like chlorine
    RTECS Number KI6300000
    Molecular Formula C2HCl5
    Linear Formula CCl3CHCl2
    Beilstein 1736845
    Melting Point −29°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PENTACHLOROETHANE
    Density 1.68g/mL
    Physical Form Liquid
    CAS 76-01-7
    Merck Index 14,7108
    Refractive Index 1.5030
    Health Hazard 1 Danger
    TSCA TSCA
    UN Number UN1669
    Health Hazard 2 GHS H Statement
    H372-H351
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Quantity 10g
    Chemical Name or Material Pentachloroethane
    Assay 96%

    GHS H Statement
    H372-H351
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.

    GHS P Statement
    P260-P281-P264-P308+P313-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    Wash thoroughly after handling.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit