Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pentacosane, ≥99.5% (GC), MilliporeSigma™ Supelco™
Pentacosane, an aliphatic hydrocarbon, is found to be a major component of volatile oil of Inula aschersoniana.
Supplier: MilliporeSigma Supelco 76493250MG
Description
PetroleumSpecifications
250 mg | |
Analytical Standard | |
MFCD00009353 | |
≥99.5% (GC) | |
52°C to 54°C; 53°C to 56°C | |
120°C - Closed Cup |
629-99-2 | |
CH3(CH2)23 CH3 | |
352.68 | |
Analytical Standard | |
169°C to 170°C (0.05 mmHg) | |
Neat |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
RTECSNumber : 211-123-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction