Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentadecafluorooctanoic acid, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
335-67-1
C8HF15O2
414.07
MFCD10567397
SNGREZUHAYWORS-UHFFFAOYSA-N
perfluorooctanoic acid, pentadecafluorooctanoic acid, perfluorocaprylic acid, pfoa, octanoic acid, pentadecafluoro, perfluoroctanoic acid, perfluoroheptanecarboxylic acid, pentadecafluoro-1-octanoic acid, pentadecafluoro-n-octanoic acid, perfluoro-n-octanoic acid
9554
CHEBI:35549
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoic acid
C(=O)(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
335-67-1
C8HF15O2
414.07
MFCD10567397
SNGREZUHAYWORS-UHFFFAOYSA-N
perfluorooctanoic acid, pentadecafluorooctanoic acid, perfluorocaprylic acid, pfoa, octanoic acid, pentadecafluoro, perfluoroctanoic acid, perfluoroheptanecarboxylic acid, pentadecafluoro-1-octanoic acid, pentadecafluoro-n-octanoic acid, perfluoro-n-octanoic acid
9554
CHEBI:35549
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoic acid
C(=O)(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
Water 4.5% max. (K.F.)
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Boiling Point 189.0°C to 192.0°C
Melting Point 53.0°C to 60.0°C
Quantity 100g
Assay Percent Range 96%
Linear Formula CF3(CF2)6CO2H
Merck Index 15, 7272
Solubility Information Solubility in water: 3.4g/L (20°C). Other solubilities: soluble in ether and ethanol
Formula Weight 414.07
Physical Form Crystalline Powder
Percent Purity 96%
Chemical Name or Material Pentadecafluorooctanoic acid
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Suspected of causing cancer.
May damage the unborn child.
May cause harm to breast-fed children.
Causes damage to organs throu

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 206-397-9