Learn More
Pentadecanolide, 98%, Thermo Scientific Chemicals
Description
Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).
Solubility
Soluble in alcohol, dipropylene glycol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 33°C to 37°C |
Density | 0.918 |
Boiling Point | 137°C (2 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.47 |
Quantity | 25 g |
Beilstein | 143710 |
Merck Index | 14,3910 |
Solubility Information | Soluble in alcohol,dipropylene glycol. Insoluble in water. |
Formula Weight | 240.39 |
Show More |
Safety and Handling
Hazard Category | H317 |
Precautionary Statement | P261-P272-P280g-P302+P352-P333+P313-P363-P501c |
DOTInformation | Hazard Class: 9; Packaging Group: III |
EINECSNumber | 203-354-6 |
RTECSNumber | RN9775000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.