Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluoro-L-phenylalanine 95.0+%, TCI America™

Click to view available options
Quantity:
200mg
1g
34702-59-5
C9H6F5NO2
255.14
MFCD01860881
YYTDJPUFAVPHQA-VKHMYHEASA-N
2761496
(2S)-2-amino-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid
N[C@@H](CC1=C(F)C(F)=C(F)C(F)=C1F)C(O)=O
34702-59-5
C9H6F5NO2
255.14
MFCD01860881
YYTDJPUFAVPHQA-VKHMYHEASA-N
2761496
(2S)-2-amino-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid
N[C@@H](CC1=C(F)C(F)=C(F)C(F)=C1F)C(O)=O
Color White-Yellow
Melting Point 258°C
Quantity 1g
Formula Weight 255.14
Physical Form Crystal-Powder at 20°C
Percent Purity ≥95.0% (HPLC,T)
Chemical Name or Material Pentafluoro-L-phenylalanine