Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorobenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
2.5g
10g
50g
Packaging:
Glass bottle
653-37-2
C7HF5O
196.08
MFCD00003303
QJXCFMJTJYCLFG-UHFFFAOYSA-N
pentafluorobenzaldehyde,perfluorobenzaldehyde,benzaldehyde, pentafluoro,benzaldehyde, 2,3,4,5,6-pentafluoro,unii-s9yie3jz4k,pentafluor-benzaldehyd,s9yie3jz4k,pubchem4253,pentafluoro-benzaldehyde,acmc-209nqh
69558
2,3,4,5,6-pentafluorobenzaldehyde
C(=O)C1=C(C(=C(C(=C1F)F)F)F)F
653-37-2
C7HF5O
196.08
MFCD00003303
QJXCFMJTJYCLFG-UHFFFAOYSA-N
pentafluorobenzaldehyde,perfluorobenzaldehyde,benzaldehyde, pentafluoro,benzaldehyde, 2,3,4,5,6-pentafluoro,unii-s9yie3jz4k,pentafluor-benzaldehyd,s9yie3jz4k,pubchem4253,pentafluoro-benzaldehyde,acmc-209nqh
69558
2,3,4,5,6-pentafluorobenzaldehyde
C(=O)C1=C(C(=C(C(=C1F)F)F)F)F
Melting Point 20°C
Boiling Point 164°C to 166°C
Flash Point 77°C
Specific Gravity 1.588
Refractive Index 1.4490 to 1.451
Infrared Spectrum Authentic
Color Yellow
Density 1.5880g/mL
Quantity 50g
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula C6F5CHO
Formula Weight 196.08
Physical Form Liquid After Melting
Percent Purity ≥97.5%
Chemical Name or Material Pentafluorobenzaldehyde
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger