Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorobenzonitrile, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
773-82-0
C7F5N
193.07
YXWJGZQOGXGSSC-UHFFFAOYSA-N
pentafluorobenzonitrile,perfluorobenzonitrile,benzonitrile, pentafluoro,pentafluorocyanobenzene,benzonitrile, 2,3,4,5,6-pentafluoro,2,3,4,5,6-pentafluoro benzonitrile,2,3,4,5,6-pentafluorobenzenecarbonitrile,cyanopentafluorobenzene,pubchem2319,acmc-209p9k
69882
2,3,4,5,6-pentafluorobenzonitrile
C(#N)C1=C(C(=C(C(=C1F)F)F)F)F
773-82-0
C7F5N
193.07
YXWJGZQOGXGSSC-UHFFFAOYSA-N
pentafluorobenzonitrile,perfluorobenzonitrile,benzonitrile, pentafluoro,pentafluorocyanobenzene,benzonitrile, 2,3,4,5,6-pentafluoro,2,3,4,5,6-pentafluoro benzonitrile,2,3,4,5,6-pentafluorobenzenecarbonitrile,cyanopentafluorobenzene,pubchem2319,acmc-209p9k
69882
2,3,4,5,6-pentafluorobenzonitrile
C(#N)C1=C(C(=C(C(=C1F)F)F)F)F
Refractive Index 1.4415 to 1.4435
Melting Point 1.0°C to 3.0°C
Infrared Spectrum Authentic
Flash Point 29°C
Quantity 5g
Density 1.5300g/mL
Specific Gravity 1.53
Boiling Point 161.0°C to 162.0°C
Assay Percent Range 98.5% min. (GC)
Linear Formula C6F5CN
Solubility Information Solubility in water: insoluble.
Formula Weight 193.07
Percent Purity 99%
Chemical Name or Material Pentafluorobenzonitrile
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

EINECSNumber : 212-259-9