Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorobenzoyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
2251-50-5
C7ClF5O
230.52
MYHOHFDYWMPGJY-UHFFFAOYSA-N
pentafluorobenzoyl chloride,perfluorobenzoyl chloride,benzoyl chloride, pentafluoro,pentafluorobenzoylchloride,pubchem15522,acmc-1covi,intermediates-zcf02601,pentafluorobenzoic acid chloride,attercop-chm at133861,67924-15-6 unspecified acetate
75256
CHEBI:39425
2,3,4,5,6-pentafluorobenzoyl chloride
C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)Cl
2251-50-5
C7ClF5O
230.52
MYHOHFDYWMPGJY-UHFFFAOYSA-N
pentafluorobenzoyl chloride,perfluorobenzoyl chloride,benzoyl chloride, pentafluoro,pentafluorobenzoylchloride,pubchem15522,acmc-1covi,intermediates-zcf02601,pentafluorobenzoic acid chloride,attercop-chm at133861,67924-15-6 unspecified acetate
75256
CHEBI:39425
2,3,4,5,6-pentafluorobenzoyl chloride
C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)Cl
Acylation reagent
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4520 to 1.4540
Specific Gravity 1.6
Color Colorless to Yellow
Quantity 5g
Density 1.6000g/mL
Assay Percent Range 97.5% min. (GC)
Linear Formula C6F5COCl
Solubility Information Solubility in water: decomposes.
Formula Weight 230.52
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Pentafluorobenzoyl chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 218-842-4