Alfa Aesar™ (Pentafluoroethyl)trimethylsilane, 97%

Manufacturer: Alfa Aesar™ H2577506

Quantity 5g

 View more versions of this product

Catalog No. AAH2577506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 124898-13-1
    Molecular Formula C5H9F5Si
    Formula Weight 192.21
    MDL Number MFCD04116436
    UN Number UN1993
    Chemical Name or Material (Pentafluoroethyl)trimethylsilane

    Specifications

    Formula Weight 192.21
    MDL Number MFCD04116436
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H224-H315-H319-H335
    Extremely flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Molecular Formula C5H9F5Si
    CAS 124898-13-1
    UN Number UN1993
    Quantity 5g
    Chemical Name or Material (Pentafluoroethyl)trimethylsilane
    Assay 97%

    Description

    Applications
    (Pentafluoroethyl)trimethylsilane acts as a silicon-based fluorinating reagent for trifluoromethylation of carbonyls and ethylene diamines. It is also employed as a precursor for metal-mediated trifluoromethylation of aromatic, heteroaromatic and vinyl substrates.

    Solubility
    Miscible with tetrahydrofuran.

    Notes
    Moisture sensitive. Incompatible with oxidizing agents.

    GHS H Statement
    H224-H315-H319-H335
    Extremely flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit