Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorophenol, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
100g
Packaging:
Glass bottle
771-61-9
C6HF5O
184.07
MFCD00002156
XBNGYFFABRKICK-UHFFFAOYSA-N
pentafluorophenol,phenol, pentafluoro,perfluorophenol,unii-a2ycf0yuha,a2ycf0yuha,2,3,4,5,6-pentafluorohydroxybenzene,c6f5oh,2,3,4,5,6-pentafluoro-phenol,phenol, 2,3,4,5,6-pentafluoro,hopfp
13041
2,3,4,5,6-pentafluorophenol
C1(=C(C(=C(C(=C1F)F)F)F)F)O
771-61-9
C6HF5O
184.07
MFCD00002156
XBNGYFFABRKICK-UHFFFAOYSA-N
pentafluorophenol,phenol, pentafluoro,perfluorophenol,unii-a2ycf0yuha,a2ycf0yuha,2,3,4,5,6-pentafluorohydroxybenzene,c6f5oh,2,3,4,5,6-pentafluoro-phenol,phenol, 2,3,4,5,6-pentafluoro,hopfp
13041
2,3,4,5,6-pentafluorophenol
C1(=C(C(=C(C(=C1F)F)F)F)F)O
Coupling reagent
Melting Point 34°C to 36°C
Boiling Point 143°C
Flash Point 72°C
Infrared Spectrum Authentic
Color White
Quantity 10g
Packaging Glass bottle
Assay Percent Range 99+%
Linear Formula C6F5OH
Merck Index 15, 7223
Solubility Information Solubility in water: soluble
Formula Weight 184.07
Physical Form Low Melting Crystalline Solid
Percent Purity ≥99.0%
Chemical Name or Material Pentafluorophenol, 99+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-235-8

RTECSNumber : SM6680000

TSCA : TSCA