Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (Pentafluorophenyl)diphenylphosphine, 98%

Manufacturer:  Alfa Aesar™ A1142206

Quantity 5g

 View more versions of this product

Catalog No. AAA1142206


  Check Availability
Add to cart

Specifications

Description

Applications
The reaction of (pentafluorophenyl)diphenylphosphine with C-ethoxycarbonyl-(p-methoxyphenyl) nitrile imine was used to synthesize [2,3,4,5-tetrafluoro-6-(p- methoxyphenylamino)phenyl]diphenylphosphine oxide whose reduction with trichlorosilane. the reactions of (pentafluorophenyl)diphenylphosphine and bis(penta- fluorophenyl)(phenyl)phosphine with nitrilimines (Housgen 1,3-dipoles) involves two-stage cycloaddi- tion and leads to formation of sufficiently stable fluorophosphoranes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

5525-95-1
352.25
Diphenylpentafluorophenylphosphine
C18H10F5P
MFCD00000290
(Pentafluorophenyl)diphenylphosphine

Specifications

5525-95-1
C18H10F5P
352.25
Diphenylpentafluorophenylphosphine
98%
70° to 72°C
5g
MFCD00000290
(Pentafluorophenyl)diphenylphosphine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit