Alfa Aesar™ (Pentafluorophenyl)diphenylphosphine, 98%

Manufacturer: Alfa Aesar™ A1142203

Quantity 1g

 View more versions of this product

Catalog No. AAA1142203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5525-95-1
    Molecular Formula C18H10F5P
    Formula Weight 352.25
    MDL Number MFCD00000290
    Synonym Diphenylpentafluorophenylphosphine
    Chemical Name or Material (Pentafluorophenyl)diphenylphosphine

    Specifications

    Formula Weight 352.25
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C18H10F5P
    Quantity 1g
    Health Hazard 1 Warning
    Melting Point 70° to 72°C
    MDL Number MFCD00000290
    CAS 5525-95-1
    Synonym Diphenylpentafluorophenylphosphine
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material (Pentafluorophenyl)diphenylphosphine
    Assay 98%

    Description

    Applications
    The reaction of (pentafluorophenyl)diphenylphosphine with C-ethoxycarbonyl-(p-methoxyphenyl) nitrile imine was used to synthesize [2,3,4,5-tetrafluoro-6-(p- methoxyphenylamino)phenyl]diphenylphosphine oxide whose reduction with trichlorosilane. the reactions of (pentafluorophenyl)diphenylphosphine and bis(penta- fluorophenyl)(phenyl)phosphine with nitrilimines (Housgen 1,3-dipoles) involves two-stage cycloaddi- tion and leads to formation of sufficiently stable fluorophosphoranes.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit